CAS RN 119313-12-1
IUPAC Navn:
2-benzyl-2-(dimethylamino)-1-[4-(morpholin-4-yl)phenyl]butan-1-one
Synonymer:
benzyl dimethylamino morpholinophenyl butanone
Molekylvægt (g/mol):
366.51
Molekylformel:
C23H30N2O2
InChi Key:
UHFFVFAKEGKNAQ-UHFFFAOYNA-N
SMILES:
CCC(CC1=CC=CC=C1)(N(C)C)C(=O)C1=CC=C(C=C1)N1CCOCC1
1
–
3
af
3
Resultater
1
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.