CAS RN 1188296-52-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
PF-4800567, MedChemExpress
MedChemExpress PF-4800567 is a potent and selective inhibitor of casein kinase 1ε (CK1ε), with an IC50 of 32 nM, which is greater than 20-fold selectivity over CK1δ (IC50, 711 nM).