CAS RN 117977-21-6
IUPAC Navn:
2-({[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methyl}sulfanyl)-1H-1,3-benzodiazole
Synonymer:
2-({[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methyl}sulfanyl)-1H-1,3-benzodiazole
Molekylvægt (g/mol):
343.45
Molekylformel:
C18H21N3O2S
InChi Key:
BSXAHDOWMOSVAP-UHFFFAOYSA-N
SMILES:
COCCCOC1=C(C)C(CSC2=NC3=CC=CC=C3N2)=NC=C1
1
–
2
af
2
Resultater
1
2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Rabeprazole Sulfide, TRC
CAS: 117977-21-6 Molekylær formel: C18 H21 N3 O2 S Molekylvægt (g/mol): 343.4432 Synonym: 2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole IUPAC navn: 2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfanyl]-1H-benzimidazole SMIL: COCCCOc1ccnc(CSc2nc3ccccc3[nH]2)c1C