Få mere at vide
2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole, Mikromol™
Artikelnummer. 30050053
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050053 25 mg
17960726 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050053 Leverandør Mikromol Leverandørnr. MM0618.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(((4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl)methyl)thio)-1H-benzimidazole
CAS 117977-21-6
Analyt- eller komponentnavne 2-(((4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl)methyl)thio)-1H-benzimidazole
Forsendelsestilstand Room Temperature
Molekylær formel C18 H21 N3 O2 S
Certificeringer/compliance ISO 17025
Synonym Rabeprazole Sodium Imp. B (EP), Rabeprazole Imp. B (EP), Rabeprazole USP Related Compound E, Rabeprazole USP RC E, 2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole, Rabeprazole Related Compound E, Rabeprazole Sodium Impurity B, Rabeprazole Impurity B
SMIL COCCCOc1ccnc(CSc2nc3ccccc3[nH]2)c1C
Molekylvægt (g/mol) 343.45
Mængde 25 mg
Formel vægt 343.445 g/mol