CAS RN 114457-94-2
IUPAC Navn:
3-phenyl-2-[(pyrazin-2-yl)formamido]propanoic acid
Synonymer:
3-phenyl-2-(pyrazin-2-ylformamido)propanoic acid
Molekylvægt (g/mol):
271.28
Molekylformel:
C14H13N3O3
InChi Key:
DWYZPDHMMZGQAP-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC=CC=C1)NC(=O)C1=NC=CN=C1
1
–
2
af
2
Resultater
1
(2S)-3-Phenyl-2-[[(pyrazin-2-yl)carbonyl]amino]propanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-Pyrazinylcarbonyl-L-phenylalanine, TRC
CAS: 114457-94-2 Molekylær formel: C14 H13 N3 O3 Molekylvægt (g/mol): 271.27 Synonym: L-Phenylalanine, N-(pyrazinylcarbonyl)- (9CI),N-(2-Pyrazinylcarbonyl)-L-phenylalanine,(S)-3-Phenyl-2-[[(pyrazin-2-yl)carbonyl]amino]propanoic acid,Bortezomib acid analog,(2S)-3-Phenyl-2-[[(pyrazin-2-yl)carbonyl]amino]propanoic Acid IUPAC navn: (2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoic acid SMIL: OC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2