Få mere at vide
N-Pyrazinylcarbonyl-L-phenylalanine, TRC
Artikelnummer. 30136007
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136007 1 g
17921825 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136007 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8415001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 114457-94-2
Molekylær formel C14 H13 N3 O3
Molekylvægt (g/mol) 271.27
Synonym L-Phenylalanine, N-(pyrazinylcarbonyl)- (9CI), N-(2-Pyrazinylcarbonyl)-L-phenylalanine, (S)-3-Phenyl-2-[[(pyrazin-2-yl)carbonyl]amino]propanoic acid, Bortezomib acid analog, (2S)-3-Phenyl-2-[[(pyrazin-2-yl)carbonyl]amino]propanoic Acid
IUPAC navn (2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoic acid
SMIL OC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2
Tekniske data
Kemisk navn eller materiale N-Pyrazinylcarbonylphenylalanine
InChI formel InChI=1S/C14H13N3O3/c18-13(12-9-15-6-7-16-12)17-11(14(19)20)8-10-4-2-1-3-5-10/h1-7,9,11H,8H2,(H,17,18)(H,19,20)/t11-/m0/s1
Mængde 1 g
Formel vægt 271.0957