CAS RN 113305-63-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2-(Phenyl-d5)-2-propanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Phenyl-d5)-2-propanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(Phenyl-d5)-2-propanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.