missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(Phenyl-d5)-2-propanol, TRC

Artikelnummer. 30422046
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422046 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422046 Leverandør Toronto Research Chemicals Leverandørnr. TRCP33604925MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Kemisk navn eller materiale 2-(Phenyl-d5)-2-propanol
CAS 113305-63-8
Molekylær formel C9H7D5O
Mængde 25 mg
Synonym 2-(Pentadeuterophenyl)-2-propanol, α,α-Dimethylbenzene-2,3,4,5,6-d5-methanol
InChI formel InChI=1S/C9H12O/c1-9(2,10)8-6-4-3-5-7-8/h3-7,10H,1-2H3/i3D,4D,5D,6D,7D
SMIL OC(C)(C)C1=C(C([2H])=C(C([2H])=C1[2H])[2H])[2H]
IUPAC navn 2-(phenyl-d5)propan-2-ol
Molekylvægt (g/mol) 141.22
Formel vægt 141.12
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.