Få mere at vide
2-(Phenyl-d5)-2-propanol, TRC
Artikelnummer. 30424161
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30424161 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424161 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3360495MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Phenyl-d5)-2-propanol
CAS 113305-63-8
Molekylær formel C9H7D5O
Mængde 5 mg
Synonym 2-(Pentadeuterophenyl)-2-propanol, α,α-Dimethylbenzene-2,3,4,5,6-d5-methanol
InChI formel InChI=1S/C9H12O/c1-9(2,10)8-6-4-3-5-7-8/h3-7,10H,1-2H3/i3D,4D,5D,6D,7D
SMIL OC(C)(C)C1=C(C([2H])=C(C([2H])=C1[2H])[2H])[2H]
IUPAC navn 2-(phenyl-d5)propan-2-ol
Molekylvægt (g/mol) 141.22
Formel vægt 141.12