CAS RN 113079-81-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-(Dimethylamino)ethyl Phenylacetate Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-(Dimethylamino)ethyl Ester Benzeneacetic Acid Hydrochloride, TRC
CAS: 113079-81-5 Molekylær formel: C12 H17 N O2 . Cl H Molekylvægt (g/mol): 243.73 Synonym: Benzeneacetic acid, 2-(dimethylamino)ethyl ester, hydrochloride (1:1),Benzeneacetic acid, 2-(dimethylamino)ethyl ester, hydrochloride (9CI),2-(Dimethylamino)ethyl Phenylacetate Hydrochloride,Cyclopentolate Hydrochloride Imp. C (EP) as Hydrochloride IUPAC navn: 2-(dimethylamino)ethyl 2-phenylacetate;hydrochloride SMIL: Cl.CN(C)CCOC(=O)Cc1ccccc1