Få mere at vide
2-(Dimethylamino)ethyl Ester Benzeneacetic Acid Hydrochloride, TRC
Artikelnummer. 30138777
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138777 500 mg
30135950 250 mg
17970365 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138777 Leverandør Toronto Research Chemicals Leverandørnr. TRCC909635500MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 113079-81-5
Molekylær formel C12 H17 N O2 . Cl H
Molekylvægt (g/mol) 243.73
Synonym Benzeneacetic acid, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Benzeneacetic acid, 2-(dimethylamino)ethyl ester, hydrochloride (9CI), 2-(Dimethylamino)ethyl Phenylacetate Hydrochloride, Cyclopentolate Hydrochloride Imp. C (EP) as Hydrochloride
IUPAC navn 2-(dimethylamino)ethyl 2-phenylacetate;hydrochloride
SMIL Cl.CN(C)CCOC(=O)Cc1ccccc1
Tekniske data
Kemisk navn eller materiale 2-(Dimethylamino)ethyl Phenylacetate Hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C12H17NO2.ClH/c1-13(2)8-9-15-12(14)10-11-6-4-3-5-7-11;/h3-7H,8-10H2,1-2H3;1H
Formel vægt 243.1026