CAS RN 1129-41-5
IUPAC Navn:
3-methylphenyl N-methylcarbamate
Synonymer:
kumiai tsumaunka MTMC tsumacide dicresyl N-methylcarbamate N-methyl-M-tolylcarbamate metholcarb metacrate metolcarb
Molekylvægt (g/mol):
165.19
Molekylformel:
C9H11NO2
InChi Key:
VOEYXMAFNDNNED-UHFFFAOYSA-N
SMILES:
CNC(=O)OC1=CC=CC(C)=C1
1
–
3
af
3
Resultater
1
Metolcarb, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Metolcarb, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Metolcarb, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.