Få mere at vide
Metolcarb, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Metolcarb |
| CAS | 1129-41-5 |
| Molekylær formel | C9 H11 N O2 |
| Mængde | 100 mg |
| Synonym | Carbamic acid, N-methyl-, 3-methylphenyl ester, Carbamic acid, methyl-, 3-methylphenyl ester (9CI), Carbamic acid, methyl-, m-tolyl ester (8CI), 3-Methylphenyl methylcarbamate, 3-Tolyl methylcarbamate, DRC 3341, MTMC, Metacrate, Metolcarb, NSC 91193, S 1065, Tsumacide, Tsumaunka, m-Cresyl N-methylcarbamate, m-Cresyl methylcarbamate, m-Methylphenyl N-methylcarbamate, m-Methylphenyl methylcarbamate, m-Tolyl methylcarbamate |
| InChI formel | InChI=1S/C9H11NO2/c1-7-4-3-5-8(6-7)12-9(11)10-2/h3-6H,1-2H3,(H,10,11) |
| SMIL | CNC(=O)Oc1cccc(C)c1 |
| IUPAC navn | (3-methylphenyl) N-methylcarbamate |
| Molekylvægt (g/mol) | 165.19 |
| Formel vægt | 165.079 |