CAS RN 1112-02-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Acetic-2,2,2-d3 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Acetic-2,2,2-d3 Acid, TRC
CAS: 1112-02-3 Molekylær formel: C2 D3 H O2 Molekylvægt (g/mol): 63.07 Synonym: Acetic Acid 2,2,2-D3,2,2,2-Trideuterioacetic acid,Acetic acid-d3,d3-Acetic acid,Trideuterioacetic acid,Acetic-d3 acid (6CI, 7CI, 8CI, 9CI) IUPAC navn: 2,2,2-trideuterioacetic acid SMIL: [2H]C([2H])([2H])C(=O)O