Få mere at vide
Acetic-2,2,2-d3 Acid, CDN
Artikelnummer. 30318108
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30318108 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318108 Leverandør CDN Leverandørnr. CDND02905G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetic-2,2,2-d3 Acid
CAS 1112-02-3
Molekylær formel C2 D3 H O2
Mængde 5 g
Synonym Acetic Acid 2,2,2-D3, 2,2,2-Trideuterioacetic acid, Acetic acid-d3, d3-Acetic acid, Trideuterioacetic acid, Acetic-d3 acid (6 CI, 7 CI, 8 CI, 9 CI)
InChI formel InChI=1 S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,2-trideuterioacetic acid
Molekylvægt (g/mol) 63.07
Formel vægt 63.04 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity