Få mere at vide
Acetic-2,2,2-d3 Acid, TRC
Artikelnummer. 30179285
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179285 500 mg
30179100 100 mg
30179206 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179285 Leverandør Toronto Research Chemicals Leverandørnr. TRCA167646500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1112-02-3
Molekylær formel C2 D3 H O2
Molekylvægt (g/mol) 63.07
Synonym Acetic Acid 2,2,2-D3, 2,2,2-Trideuterioacetic acid, Acetic acid-d3, d3-Acetic acid, Trideuterioacetic acid, Acetic-d3 acid (6CI, 7CI, 8CI, 9CI)
IUPAC navn 2,2,2-trideuterioacetic acid
SMIL [2H]C([2H])([2H])C(=O)O
Tekniske data
Kemisk navn eller materiale Acetic-2,2,2-d3 Acid
Mængde 500 mg
InChI formel InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3
Formel vægt 63.04