Få mere at vide
ZYZ-488, MedChemExpress
Artikelnummer. 16494336
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16494336 100 mg
16443038 10 mg
16419606 50 mg
16415777 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16494336 Leverandør Medchem Express Leverandørnr. HY100472
MedChemExpress ZYZ-488 is a competitive apoptotic protease activating factor-1 (Apaf-1) inhibitor. ZYZ-488 inhibits the activation of binding protein procaspase-9 and procaspase-3.
ZYZ-488 is a competitive apoptotic protease activating factor-1 (Apaf-1) inhibitor. ZYZ-488 inhibits the activation of binding protein procaspase-9 and procaspase-3.
Tekniske data
Kemisk navn eller materiale ZYZ-488
CAS 1470302-79-4
Farve White
Renhedsgrad noter Research
Molekylær formel C20H29N3O11
Mængde 100 mg
Opløselighedsinformation DMSO : 200 mg/mL (410.29 mM; Need ultrasonic)
SMIL O=C(OCCCCNC(N)=N)C(C=C1OC)=CC(OC)=C1O[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O2
Molekylvægt (g/mol) 487.46
Formel vægt 487.46
Procent renhed 99.69%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death