Learn More
Labcon ZAP™ 1000 μL Extended Length Aerosol Filter Pipette Tips
Fits Most Popular 1000 μL Pipettors
Brand: Labcon 1057-960-016-9
67.12 DKK valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 11.43060kg
Description
- Pipette Tip
- Fits Most Popular 1000 μL Pipettors
- Patented robotically inserted endotoxin free filter
- Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polypropylene USP Class VI resin
- Made in USA
Specifications
1000 to 1300 μL | |
Sterile | |
Not Autoclavable | |
88.9 mm | |
Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) | |
Rack | |
96 tips/rack; 6 racks/Pk.; 8 Pk./case | |
Single-use | |
No | |
Polypropylene Resin |
Filtered | |
Natural | |
3.50 in. | |
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity | |
Liquid Handing | |
UltraFine | |
96 tips | |
Universal Fit | |
ZAP |