Få mere at vide
(Z)-Cinnarizine, TRC
Artikelnummer. 17923905
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17923905 100 mg
17972845 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17923905 Leverandør Toronto Research Chemicals Leverandørnr. TRCC465290100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 750512-44-8
Molekylær formel C26 H28 N2
Molekylvægt (g/mol) 368.51
Synonym (Z)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine, Piperazine, 1-(diphenylmethyl)-4-[(2Z)-3-phenyl-2-propen-1-yl]-, Piperazine, 1-(diphenylmethyl)-4-(3-phenyl-2-propenyl)-, (Z)- (9CI), 1-(Diphenylmethyl)-4-[(2Z)-3-phenyl-2-propen-1-yl]piperazine, (Z)-Cinnarizine
IUPAC navn 1-benzhydryl-4-[(Z)-3-phenylprop-2-enyl]piperazine
SMIL C(\C=C/c1ccccc1)N2CCN(CC2)C(c3ccccc3)c4ccccc4
Tekniske data
Kemisk navn eller materiale (Z)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C26H28N2/c1-4-11-23(12-5-1)13-10-18-27-19-21-28(22-20-27)26(24-14-6-2-7-15-24)25-16-8-3-9-17-25/h1-17,26H,18-22H2/b13-10-
Formel vægt 368.2252
Procent renhed >95%