Learn More
Invitrogen™ Human YES1, GST Tag Recombinant Protein
YES1 is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
3925.00 DKK - 56200.00 DKK
Specifications
Accession Number | NP_005424 |
---|---|
Gene Alias | YES1 |
KinaseFamily | SRC Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15313816
|
Invitrogen™
A15557 |
10μg |
3925.00 DKK
10µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15323816
|
Invitrogen™
A15558 |
100μg |
13020.00 DKK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15333816
|
Invitrogen™
A15559 |
1mg |
56200.00 DKK
1mg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Active—this protein can be used in kinase activity assays, such as Z’-LYTE™ assays, or in a binding assay platform like the LanthaScreen™ Eu Kinase Binding Assay (KBA).
- Purified—purity is determined by gel electrophoresis and provided in lot-specific certificate of analysis (COA).
- GST-tagged—provides animal origin flexibility. When used in the KBA platform, it can be paired with a goat- or rabbit-derived Eu-anti-GST antibody.
Specifications
NP_005424 | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |
YES1 | |
Human | |
GST Tag |