Learn More
Invitrogen™ Human YES1, GST Tag Recombinant Protein
YES1 is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
Brand: Invitrogen™ A15558
Additional Details : Weight : 0.10000kg
Description
- Active—this protein can be used in kinase activity assays, such as Z’-LYTE™ assays, or in a binding assay platform like the LanthaScreen™ Eu Kinase Binding Assay (KBA).
- Purified—purity is determined by gel electrophoresis and provided in lot-specific certificate of analysis (COA).
- GST-tagged—provides animal origin flexibility. When used in the KBA platform, it can be paired with a goat- or rabbit-derived Eu-anti-GST antibody.
Specifications
NP_005424 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
100μg | |
RUO - research use only | |
YES1 | |
Human | |
GST Tag | |
Frozen |
Protein Biology, Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis | |
88.3kDa | |
For maximum recovery please spin prior to use. Aliquoting the 5μg, 10μg, and 20μg sizes of kinase is not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 , μL are not recommended. Never store a kinase diluted. If properly stored at -80°C, this product is guaranteed for 6 months from date of purchase. | |
Full Length | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |