Få mere at vide
WZ4002, MedChemExpress
Artikelnummer. 16474928
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16474928 10 mg
16441177 50 mg
16465706 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16474928 Leverandør Medchem Express Leverandørnr. HY12026
MedChemExpress WZ4002 is a mutant selective EGFR inhibitor with IC50s of 2, 8, 3 and 2 nM for EGFRL858R, EGFRL858R/T790M, EGFRE746_A750 and EGFRE746_A750/T790M, respectively.
WZ4002 is a mutant selective EGFR inhibitor with IC50s of 2, 8, 3 and 2 nM for EGFRL858R, EGFRL858R/T790M, EGFRE746_A750 and EGFRE746_A750/T790M, respectively.
Tekniske data
Kemisk navn eller materiale WZ4002
CAS 1213269-23-8
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C25H27ClN6O3
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 100 mg/mL (202.03 mM)
SMIL COC1=C(C=CC(N2CCN(CC2)C)=C1)NC3=NC=C(C(OC4=CC(NC(C=C)=O)=CC=C4)=N3)Cl
Molekylvægt (g/mol) 494.97
Formel vægt 494.97
Procent renhed 99.69%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease