Learn More
Bohlender™ Within Luer Lock PTFE Miniature Luer-Lock Connector
Connect componets with Luer Lock such as syringes or cannulas to a tubing system.
Brand:  Bohlender™ F718-04
 View more versions of this product
Product Code. 13277169
  • 162.00 DKK / Each
Only null left
Estimated Shipment: 17-07-2025
Description
  • Connector made of PTFE
  • For transition from Luer Lock connection to female thread UNF 1/4 in. 28G
  • Inner cone of the Luer-Lock connection provides good sealing, the additional thread (either male or female) prevents accidental loosening
  • Tubing is connected by means of tube end fittings to a female thread UNF 1/4 in. 28G
Specifications
Specifications
1.6 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bohlender™ Within Luer Lock PTFE Miniature Luer-Lock Connector > I.D.: 1.6mm