Learn More
Wheaton™ Shaker Flasks With Four Baffles
Autoclavable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Brand:  Wheaton™ 354244
678.84 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15297534
  • 873.00 DKK / Each
Only null left
Estimated Shipment: 21-08-2025
Description
  • For use with orbital or reciprocating shakers
  • Baffles improve aeration and mixing for microbial and bacterial cultures
  • Break resistant lip for use with stopper, cotton plug or film cover
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Wheaton™ Shaker Flasks With Four Baffles > 1000mL