Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE Faced 14B White Rubber Liner
Provides completely inert inner seal and surface for sample or product
Brand: DWK Life Sciences W240406
1814.06 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Autoclavable
Specifications
8-425 | |
Screw Cap | |
Phenolic |
TFE, Styrene Butadiene Rubber | |
Autoclavable | |
Black |
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE Faced 14B White Rubber Liner > Screw cap size: 8mm-425
Spot an opportunity for improvement?Share a Content Correction