Learn More
Welch™ Vacuum Pump On/Off Foot Switch
Vacuum pump accessory that provides hands-free operation of Welch DryFast diaphragm and WOB-L piston pumps.
1878.94 DKK
Product Code | Brand | For Use With | Electrical Requirements | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Electrical Requirements | Model | Price | Quantity & Availability | |||
10349442
|
Welch™
1430C |
WOB-L™ piston pumps | 90 to 230 V | Momentary On/Off switch logic |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Hands-free on/off pump operation
- No adaptors or special connection needed
- Plugs directly into the power source outlet
- Switch logic and power varies depending on model:
- Parts 01-104-17 and 01-055-16: 3-prong U.S. standard plug with 8 ft. cord (CSA, NEMA, and UL Enclosure Type 1)
- Part 01-055-17: Male and female IEC connections with 8 ft. cord (plugs directly into the IEC connection on the pump)
Specifications
Foot Switch |