missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
WDR47 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29364-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The WDR47 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WDR47. This antibody recognizes human antigen. The WDR47 antibody has been validated for the following applications: WB, ELISA analysis.Tekniske data
| WDR47 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 22911 | |
| Store at -20°C. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 900 μg/mL | |
| FLJ90135; KIAA0893 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 100 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion