Få mere at vide
VLX1570, MedChemExpress
Artikelnummer. 16475188
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16475188 10 mg
16471407 50 mg
16461259 1 mg
16427917 5 mg
16415886 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16475188 Leverandør Medchem Express Leverandørnr. HY12471
MedChemExpress VLX1570 is a competitive inhibitor of proteasome deubiquitinases (DUBs) with an IC50 of approximate 10 μM.
VLX1570 is a competitive inhibitor of proteasome deubiquitinases (DUBs) with an IC50 of approximate 10 μM.
Tekniske data
Kemisk navn eller materiale VLX1570
CAS 1431280-51-1
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C23H17F2N3O6
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 32 mg/mL (68.17 mM)
SMIL O=C1/C(CN(C(C=C)=O)CC/C1=C\C2=CC=C(F)C([N+]([O-])=O)=C2)=C/C3=CC=C(F)C([N+]([O-])=O)=C3
Molekylvægt (g/mol) 469.39
Formel vægt 469.39
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death