Beskrivelse
- Clear.
- Technology approved for continuous wear and certified with perfect optical quality.
- Lens protection from medium energy impact, resists a 6mm, 0.86g ball at 120m/s.
- CE European standards certificate available.
Tekniske data
Tekniske data
| Certificeringer/compliance | CE European standards certificate available. |
| Linsefarve | Clear |
| Produkttype | Faceshield |
| Linse materiale | Polycarbonate |