Learn More
Sartorius Stedim Biotech™ Viscose Non-Woven Filters, Discs, Grade 2601
Generally used for viscous liquid filtration in environments such as the petrochemical industry.
Brand: Sartorius Stedim Biotech™ FT-3-01304-025
Additional Details : Weight : 0.09300kg
Description
- Non-woven
- Suitable for filtration of viscous solutions.
- Application example: Petrochemical industry: Filtration of viscous solutions to retain the biggest particles before further analysis with a viscosimetry.
- Non-sterile, bulk packed.
Specifications
60 g/m2 | |
Rayon (Viscose) | |
2601 | |
0.46 mm | |
25 mm |
Disc | |
50 μm | |
100/Pk. | |
Non-woven Filter |