Få mere at vide
Vinylcyclopropane, TRC
Artikelnummer. 30450930
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450930 1 g
30456725 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30450930 Leverandør Toronto Research Chemicals Leverandørnr. TRCV3800001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 693-86-7
Molekylær formel C5 H8
Molekylvægt (g/mol) 68.12
Synonym Ethenyl-cyclopropane, Vinyl-cyclopropane, Cyclopropylethene, Cyclopropylethylene, Cyclopropyl-ethene
IUPAC navn ethenylcyclopropane
SMIL C=CC1CC1
Tekniske data
Kemisk navn eller materiale Vinylcyclopropane
Mængde 1 g
InChI formel InChI=1S/C5H8/c1-2-5-3-4-5/h2,5H,1,3-4H2
Formel vægt 68.0626