missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
VEGFR2/KDR/Flk-1 Antibody (4H3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
1250.00 DKK - 3820.00 DKK
Tekniske data
| Antigen | VEGFR2/KDR/Flk-1 |
|---|---|
| Klon | 4H3 |
| Koncentration | LYOPH |
| Applikationer | Immunofluorescence, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA, Western Blot |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18291845
|
Novus Biologicals
NBP1-18644-0.02MG |
0.02 mg |
1250.00 DKK
0.02mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18778033
|
Novus Biologicals
NBP1-18644 |
0.1 mg |
3820.00 DKK
0.10mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Tekniske data
| VEGFR2/KDR/Flk-1 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2 | |
| KDR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 4H3 | |
| Immunofluorescence, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases | |
| P35968 | |
| 3791 | |
| IgG1 | |
| Protein G purified | |
| The monoclonal antibody will detect native human VEGFR-2/KDR on the surface of different human cell types. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel