missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
VEGFR2/KDR/Flk-1 Antibody (4H3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-18644-0.02MG
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Tekniske data
| VEGFR2/KDR/Flk-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 2 - 5 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 1-5 ug/ml, CyTOF-ready | |
| P35968 | |
| KDR | |
| human recombinant CNTF | |
| 0.02 mg | |
| Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases | |
| 3791 | |
| Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunofluorescence, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA, Western Blot | |
| 4H3 | |
| Unconjugated | |
| PBS with No Preservative | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The monoclonal antibody will detect native human VEGFR-2/KDR on the surface of different human cell types. | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion