Få mere at vide
Valdecoxib Sulfonic Acid, TRC
Artikelnummer. 30180297
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180297 25 mg
30180377 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30180297 Leverandør Toronto Research Chemicals Leverandørnr. TRCV09001025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 181696-35-5
Molekylær formel C16 H13 N O4 S
Molekylvægt (g/mol) 315.34
Synonym 4-(5-Methyl-3-phenyl-4-isoxazolyl)benzenesulfonic acid
IUPAC navn 4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonic acid
SMIL Cc1onc(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Valdecoxib Sulfonic Acid
Mængde 25 mg
InChI formel InChI=1S/C16H13NO4S/c1-11-15(12-7-9-14(10-8-12)22(18,19)20)16(17-21-11)13-5-3-2-4-6-13/h2-10H,1H3,(H,18,19,20)
Formel vægt 315.0565