Learn More
BRAND™ USP Measuring Flask
With individual USP conformity certificate
Brand: BRAND™ 956938
331.64 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
5 mL | |
Borosilicate Glass (3.3) | |
Ground Joint NS 10/19 | |
A |
Product Content Correction
Product Title
BRAND™ USP Measuring Flask > Capacity (Metric): 5 mL
Spot an opportunity for improvement?Share a Content Correction