Få mere at vide
Thermo Scientific™ Urapidil hydrochloride, 98+%
alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist
Brand: Thermo Scientific™ J63219.03
| Mængde | 1 g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| 64887-14-5 | |
| 6-(3-[4-(2-Methoxypenyl)-1-piperaziny]propylamino)-1,3-dimethyluracil hydrochloride | |
| alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist | |
| ≥98% |
| 423.93g/mol | |
| Powder | |
| White |
Tekniske data
| 1 g | |
| Powder | |
| 64887-14-5 | |
| 423.93g/mol | |
| ≥98% |
| White | |
| alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist | |
| C20H29N5O3·HCl | |
| 6-(3-[4-(2-Methoxypenyl)-1-piperaziny]propylamino)-1,3-dimethyluracil hydrochloride |