Få mere at vide
UPF 1069, MedChemExpress
Artikelnummer. 16476078
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16476078 10 mg
16462217 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16476078 Leverandør Medchem Express Leverandørnr. HY14478
MedChemExpress UPF 1069 is a PARP inhibitor, with IC50s of 8 and 0.3 μM for PARP-1 and PARP-2, respectively.
UPF 1069 is a PARP inhibitor, with IC50s of 8 and 0.3 μM for PARP-1 and PARP-2, respectively.
Tekniske data
Kemisk navn eller materiale UPF 1069
CAS 1048371-03-4
Farve White
Renhedsgrad noter Research
Molekylær formel C17H13NO3
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 100 mg/mL (358.05 mM)
SMIL O=C1NC=CC2=C1C=CC=C2OCC(C3=CC=CC=C3)=O
Molekylvægt (g/mol) 279.29
Formel vægt 279.29
Procent renhed 99.2%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death