Få mere at vide
UNC0642, MedChemExpress
Artikelnummer. 16466376
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16466376 100 mg -
16445948 10 mg 10mg
16442097 50 mg -
16438577 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16466376 Leverandør Medchem Express Leverandørnr. HY13980
MedChemExpress UNC0642 is a potent and selective lysine methyltransferases G9a and GLP inhibitor, with an IC50 of G9a.
UNC0642 is a potent and selective lysine methyltransferases G9a and GLP inhibitor, with an IC50 of G9a.
Tekniske data
Kemisk navn eller materiale UNC0642
CAS 1481677-78-4
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C29H44F2N6O2
Mængde 100 mg
Opløselighedsinformation DMSO : 50 mg/mL (91.46 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL CC(N1CCC(NC2=C3C=C(OC)C(OCCCN4CCCC4)=CC3=NC(N5CCC(F)(F)CC5)=N2)CC1)C
Molekylvægt (g/mol) 546.7
Formel vægt 546.7
Procent renhed 95.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease