missing translation for 'onlineSavingsMsg'
Få mere at vide

Umbralisib hydrochloride, MedChemExpress

Artikelnummer. 16477777
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16477777 5 mg
16455796 100 mg
16445058 10 mg
16421297 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16477777 Leverandør Medchem Express Leverandørnr. HY12279C

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Umbralisib (TGR-1202) hydrochloride is an orally active, potent and selective dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibitor, with EC50 of 22.2 nM and 6.0 μM, respectively. Umbralisib hydrochloride exhibits unique immunomodulatory effects on chronic lymphocytic leukemia (CLL) T cells. Umbralisib hydrochloride can be used for haematological malignancies reseach.

Umbralisib (TGR-1202) hydrochloride is an orally active, potent and selective dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibitor, with EC50 of 22.2 nM and 6.0 μM, respectively. Umbralisib hydrochloride exhibits unique immunomodulatory effects on chronic lymphocytic leukemia (CLL) T cells. Umbralisib hydrochloride can be used for haematological malignancies reseach.

Tekniske data

Kemisk navn eller materiale Umbralisib hydrochloride
CAS 1532533-78-0
Farve White
Renhedsgrad noter Research
Molekylær formel C31H25ClF3N5O3
Mængde 5 mg
Synonym TGR-1202 hydrochloride RP5264 hydrochloride
Opløselighedsinformation DMSO : ≥ 150 mg/mL (246.71 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL O=C1C(C2=CC=CC(F)=C2)=C([C@@H](N3N=C(C4=CC=C(OC(C)C)C(F)=C4)C5=C(N)N=CN=C53)C)OC6=CC=C(F)C=C16.[H]Cl
Molekylvægt (g/mol) 608.01
Formel vægt 608.01
Procent renhed 98.98%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.