Få mere at vide
Tyvelose, TRC
Artikelnummer. 30450937
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450937 10 mg
30456453 25 mg
30454410 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30450937 Leverandør Toronto Research Chemicals Leverandørnr. TRCT94800010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5658-12-8
Molekylær formel C6H12O4
Molekylvægt (g/mol) 148.16
Synonym 3,6-Dideoxy-D-arabino-hexose, 3,6-Dideoxy-D-mannose, D-Tyvelose
IUPAC navn (3S,5S,6R)-6-methyloxane-2,3,5-triol
SMIL C[C@H]1OC(O)[C@@H](O)C[C@@H]1O
Tekniske data
Kemisk navn eller materiale Tyvelose
Mængde 10 mg
InChI formel InChI=1S/C6H12O4/c1-3-4(7)2-5(8)6(9)10-3/h3-9H,2H2,1H3/t3-,4+,5+,6?/m1/s1
Formel vægt 148.0736