Få mere at vide
TYR-PRO TFA Salt, TRC
Artikelnummer. 30450776
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450776 25 mg
30455389 5 mg
30455103 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30450776 Leverandør Toronto Research Chemicals Leverandørnr. TRCT96640525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51871-48-8
Molekylær formel C14H18N2O4 . C2HF3O2
Molekylvægt (g/mol) 392.33
Synonym L-Proline, 1-L-tyrosyl-, mono(trifluoroacetate) (salt) (9CI), Acetic acid, trifluoro-, compd. with 1-L-tyrosyl-L-proline (1:1) (ZCI), L-Tyrosyl-L-proline trifluoroacetate, (2S)-1-[(2S)-2-Amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidine-2-carboxylic acid trifluoroacetate
IUPAC navn (2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidine-2-carboxylic acid;2,2,2-trifluoroacetic acid
SMIL N[C@@H](Cc1ccc(O)cc1)C(=O)N2CCC[C@H]2C(=O)O.OC(=O)C(F)(F)F
Tekniske data
Kemisk navn eller materiale TYR-PRO TFA Salt
InChI formel InChI=1S/C14H18N2O4.C2HF3O2/c15-11(8-9-3-5-10(17)6-4-9)13(18)16-7-1-2-12(16)14(19)20;3-2(4,5)1(6)7/h3-6,11-12,17H,1-2,7-8,15H2,(H,19,20);(H,6,7)/t11-,12-;/m0./s1
Mængde 25 mg
Formel vægt 392.1195