Få mere at vide
Tucidinostat, MedChemExpress
Artikelnummer. 16483006
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
25mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16483006 25 mg 25mg
16455186 100 mg -
16450587 50 mg -
16446977 5 mg 5mg
16434258 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16483006 Leverandør Medchem Express Leverandørnr. HY109015
MedChemExpress Tucidinostat (Chidamide) is a potent and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor, with IC50s of 95, 160, 67 and 78 nM, less active on HDAC8 and HDAC11 (IC50s, 733 nM, 432 nM, respectively), and shows no effect on HDAC4/5/6/7/9.
Tucidinostat (Chidamide) is a potent and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor, with IC50s of 95, 160, 67 and 78 nM, less active on HDAC8 and HDAC11 (IC50s, 733 nM, 432 nM, respectively), and shows no effect on HDAC4/5/6/7/9.
Tekniske data
Kemisk navn eller materiale Tucidinostat
CAS 1616493-44-7
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C22H19FN4O2
Mængde 25 mg
Synonym Chidamide HBI-8000 CS 055
Opløselighedsinformation DMSO : ≥ 50 mg/mL (128.07 mM)
SMIL O=C(C1=CC=C(CNC(/C=C/C2=CN=CC=C2)=O)C=C1)NC(C=CC(F)=C3)=C3N
Molekylvægt (g/mol) 390.41
Formel vægt 390.41
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death