Learn More
Ebro Electronic™ TTX 100 Core Thermometer Thermocouple Type T
Provides for fast measurements with high accuracy. Ebro Electronic™ TTX 100 Core Thermometer Thermocouple Type T comes with a cable probe.
Brand: Ebro Electronic™ 1340-5100
This item is not returnable.
View return policy
Description
- HACCP
- Large display
- Robust and impact-resistant
Specifications
CE DIN EN 13485 | |
Thermometer to measure ambient or heart | |
90 mm | |
ABS Plastic | |
Permanently attached pointed | |
Core Thermocouple Thermometer | |
-50°C | |
±0.8°C |
3 mm | |
17 mm | |
105 mm | |
TTX 100 | |
0.1°C from −60 to 199.9°C, 1°C above | |
350°C | |
-50°C to +350°C | |
42 mm |
Product Content Correction
Product Title
Ebro Electronic™ TTX 100 Core Thermometer Thermocouple Type T > Temperature Range: -50 to 350deg.C
Spot an opportunity for improvement?Share a Content Correction