missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TTLL12 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68044-1-IG-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The TTLL12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TTLL12. This antibody recognizes human antigen. The TTLL12 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Tekniske data
| TTLL12 | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ41795; KIAA0153; dJ526I14.2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 72-74 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1H5C11 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 23170 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion