missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TRPM7 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55251-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The TRPM7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human TRPM7. This antibody recognizes human, mouse, rat antigen. The TRPM7 antibody has been validated for the following applications: WB, ELISA analysis.Tekniske data
| TRPM7 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54822 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 700 μg/mL | |
| CHAK; CHAK1; FLJ20117; FLJ25718; LTRPC7; TRP-PLIK | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 220 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion