Få mere at vide
Tripropylene Glycol Diacrylate, Mixture of Isomers, TRC
Artikelnummer. 17952395
Skift visning
Mængde:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17952395 25 g
17915035 100 g
17913035 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17952395 Leverandør Toronto Research Chemicals Leverandørnr. TRCT81401525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 42978-66-5
Molekylær formel C15H24O6
Molekylvægt (g/mol) 300.35
Synonym 310HP, APG 200, Actilane 424, AgiSyn 2815, Aronix M 220, Chemlink 51, Cognis 4061, Doublemer TPGDA ;EB 1110, EM 223, Ebecryl TPGDA, Ebecryl TRPGDA-DEO, Etermer EM 223, EvergGlide UV 636, Everglide UV 636, Genomer 1234, KS-TPGDA, Kayarad TPGDA, LM 259, Laromer TPGDA, M 220;
IUPAC navn (propane-1,3-diylbis(oxy))bis(propane-3,1-diyl) diacrylate
SMIL C=CC(OCCCOCCCOCCCOC(C=C)=O)=O
Tekniske data
Kemisk navn eller materiale Tripropylene Glycol Diacrylate, Mixture of Isomers
Mængde 25 g
InChI formel InChI=1S/C15H24O6/c1-3-14(16)20-12-6-10-18-8-5-9-19-11-7-13-21-15(17)4-2/h3-4H,1-2,5-13H2
Formel vægt 300.16