CAS RN 42978-66-5
IUPAC Navn:
1-({1-[2-(prop-2-enoyloxy)propoxy]propan-2-yl}oxy)propan-2-yl prop-2-enoate
Synonymer:
tripropylene glycol diacrylate
Molekylvægt (g/mol):
300.35
Molekylformel:
C15H24O6
InChi Key:
ZDQNWDNMNKSMHI-UHFFFAOYNA-N
SMILES:
CC(COCC(C)OC(=O)C=C)OCC(C)OC(=O)C=C
1
–
1
af
1
Resultater
1
Tripropylene Glycol Diacrylate, Mixture of Isomers, TRC
CAS: 42978-66-5 Molekylær formel: C15H24O6 Molekylvægt (g/mol): 300.35 Synonym: 310HP,APG 200,Actilane 424,AgiSyn 2815,Aronix M 220,Chemlink 51,Cognis 4061,Doublemer TPGDA ;EB 1110,EM 223,Ebecryl TPGDA,Ebecryl TRPGDA-DEO,Etermer EM 223,EvergGlide UV 636,Everglide UV 636,Genomer 1234,KS-TPGDA,Kayarad TPGDA,LM 259,Laromer TPGDA,M 220; IUPAC navn: (propane-1,3-diylbis(oxy))bis(propane-3,1-diyl) diacrylate SMIL: C=CC(OCCCOCCCOCCCOC(C=C)=O)=O