Få mere at vide
Trifluoroiodomethane, 97+%, Thermo Scientific™
2626.90 DKK
Kemiske identifikatorer
| CAS | 2314-97-8 |
|---|---|
| Molekylær formel | CF3I |
| Molekylvægt (g/mol) | 195.911 |
| MDL nummer | MFCD00001060 |
| InChI nøgle | VPAYJEUHKVESSD-UHFFFAOYSA-N |
| Synonym | trifluoromethyl iodide, iodotrifluoromethane, methane, trifluoroiodo, perfluoromethyl iodide, trifluoro iodo methane, trifluoromethyliodide, monoiodotrifluoromethane, freon 13t1, unii-42a379kb0u, ccris 7782 |
| PubChem CID | 16843 |
| IUPAC navn | trifluoro(iodo)methane |
| SMIL | C(F)(F)(F)I |
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| CF3I | |
| MFCD00001060 | |
| trifluoromethyl iodide, iodotrifluoromethane, methane, trifluoroiodo, perfluoromethyl iodide, trifluoro iodo methane, trifluoromethyliodide, monoiodotrifluoromethane, freon 13t1, unii-42a379kb0u, ccris 7782 | |
| trifluoro(iodo)methane |
Tekniske data
| Trifluoroiodomethane | |
| 2.361 g/mL | |
| None | |
| CF3I | |
| MFCD00001060 | |
| 1732740 | |
| trifluoromethyl iodide, iodotrifluoromethane, methane, trifluoroiodo, perfluoromethyl iodide, trifluoro iodo methane, trifluoromethyliodide, monoiodotrifluoromethane, freon 13t1, unii-42a379kb0u, ccris 7782 | |
| VPAYJEUHKVESSD-UHFFFAOYSA-N | |
| trifluoro(iodo)methane | |
| 16843 | |
| ≥97% |
| 2314-97-8 | |
| -22.5°C | |
| Cylinder | |
| 1.379 | |
| UN1956 | |
| Light sensitive | |
| Slightly soluble in water. | |
| C(F)(F)(F)I | |
| 195.911 | |
| 195.91 |
Sikkerhed og håndtering
GHS H Statement
H280-H341
Contains gas under pressure
may explode if heated.
Suspected of causing genetic defects.
P201-P202-P281-P308+P313-P501c
H341
missing translation for 'dotInformation' : Transport Hazard Class: 2.2; Proper Shipping Name: COMPRESSED GAS N.O.S.
missing translation for 'einecsNumber' : 219-014-5
missing translation for 'rtecsNumber' : PB6975000
missing translation for 'tsca' : Yes
Anbefalet opbevaring : Ambient temperatures
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.