missing translation for 'onlineSavingsMsg'
Få mere at vide

Triflumizole, TRC

Artikelnummer. 30040787
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
2.5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30040787 250 mg
30012359 10 g
30010633 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30040787 Leverandør Toronto Research Chemicals Leverandørnr. TRCT779150250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 68694-11-1
Molekylær formel C15 H15 Cl F3 N3 O
Molekylvægt (g/mol) 345.75
Synonym [N(E)]-4-Chloro-N-[1-(1H-imidazol-1-yl)-2-propoxyethylidene]-2-(trifluoromethyl)benzenamine, 1-[(1E)-1-[[4-Chloro-2-(trifluoromethyl)phenyl]imino]-2-propoxyethyl]-1H-imidazole, (E)-1-[1-[[4-Chloro-2-(trifluoromethyl)phenyl]imino]-2-propoxyethyl]-1H-imidazole, Condor, NF 114, Procure, Procure (fungicide), SSP 11, Terraguard, Triflumizol, Triflumizole, Trifmine, Trifumin, Trifumine
IUPAC navn (E)-N-[4-chloro-2-(trifluoromethyl)phenyl]-1-imidazol-1-yl-2-propoxyethanimine
SMIL CCCOC\C(=N/c1ccc(Cl)cc1C(F)(F)F)\n2ccnc2

Specifications

Kemisk navn eller materiale Triflumizole
Mængde 250 mg
InChI formel InChI=1S/C15H15ClF3N3O/c1-2-7-23-9-14(22-6-5-20-10-22)21-13-4-3-11(16)8-12(13)15(17,18)19/h3-6,8,10H,2,7,9H2,1H3/b21-14+
Formel vægt 345.0856
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.