Få mere at vide
Triethyloxonium Tetrachloroferrate(III), TRC
Artikelnummer. 30450899
Skift visning
Mængde:
2.5 g
5 g
10 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30450899 5 g -
30454948 10 g -
30455926 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30450899 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7769055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15555-35-8
Molekylær formel C6H15Cl4FeO
Molekylvægt (g/mol) 300.84
Synonym Triethyloxonium Chloroferrate, Triethyloxonium Tetrachloroferrate
IUPAC navn tetrachloroiron(1+);triethyloxidanium
SMIL CC[O+](CC)CC.Cl[Fe+](Cl)(Cl)Cl
Tekniske data
Kemisk navn eller materiale Triethyloxonium Tetrachloroferrate(III)
Mængde 5 g
InChI formel InChI=1S/C6H15O.4ClH.Fe/c1-4-7(5-2)6-3;;;;;/h4-6H2,1-3H3;4*1H;/q+1;;;;;+5/p-4
Formel vægt 298.9226